Masterflex® Male Luer to Barb Fitting, Avantor®
Supplier: VWR International
MFLX40318-00 MFLX40318-01 MFLX40318-02 MFLX40318-03 MFLX40318-04 MFLX40318-05 MFLX40318-06 MFLX40318-07 MFLX40318-08 MFLX40318-09 MFLX40318-10 MFLX40318-11 MFLX40318-12 MFLX40318-13 MFLX40318-14 MFLX40318-15 MFLX40318-16 MFLX40318-17 MFLX40318-18 MFLX40318-19 MFLX40318-20 MFLX40318-21 MFLX40318-22 MFLX40318-23 MFLX40318-24 MFLX40318-25 MFLX40318-26 MFLX40318-27 MFLX40318-28 MFLX40318-29
MFLX40318-00EA 17.38 CAD
MFLX40318-00 MFLX40318-01 MFLX40318-02 MFLX40318-03 MFLX40318-04 MFLX40318-05 MFLX40318-06 MFLX40318-07 MFLX40318-08 MFLX40318-09 MFLX40318-10 MFLX40318-11 MFLX40318-12 MFLX40318-13 MFLX40318-14 MFLX40318-15 MFLX40318-16 MFLX40318-17 MFLX40318-18 MFLX40318-19 MFLX40318-20 MFLX40318-21 MFLX40318-22 MFLX40318-23 MFLX40318-24 MFLX40318-25 MFLX40318-26 MFLX40318-27 MFLX40318-28 MFLX40318-29
Masterflex® Male Luer to Barb Fitting, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information