Masterflex® Barbed Low Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: VWR International
MFLX40251-00 MFLX40250-99 MFLX40250-92 MFLX40250-91 MFLX40250-85 MFLX40250-84 MFLX40250-77 MFLX40250-76 MFLX40250-69 MFLX40250-68 MFLX40250-61 MFLX40250-60
MFLX40251-00EA 181.7 CAD
MFLX40251-00 MFLX40250-99 MFLX40250-92 MFLX40250-91 MFLX40250-85 MFLX40250-84 MFLX40250-77 MFLX40250-76 MFLX40250-69 MFLX40250-68 MFLX40250-61 MFLX40250-60
Masterflex® Barbed Low Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Low Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information