Seametrics Low-Flow Impeller Sensors
Supplier: SEAMETRICS INC
SPX-038 SPX-050 SPX-100 MFLX33110-05 MFLX33110-20 MFLX33110-25 MFLX33110-35 MFLX33125-10 MFLX33125-15 MFLX33125-20
MFLX33110-00EA 677.82 CAD
MFLX33110-00 MFLX33110-10 MFLX33110-30 MFLX33110-05 MFLX33110-20 MFLX33110-25 MFLX33110-35 MFLX33125-10 MFLX33125-15 MFLX33125-20
Seametrics Low-Flow Impeller Sensors
Flow Meters
Impeller design minimizes wear for long sensor life.
  • Utilizes rotating turbine and a magnetic Hall-effect sensors
Each flow sensor uses a rotating turbine and a magnetic Hall-effect sensor to generate an electronic pulse proportional to flow rate. The controller converts these pulses into flow units. These low-flow sensors measure flow rates in pipe sizes from 3/8 to 1" NPT(F). Sensors generate a square wave pulse output that can be sent to flow controllers 33110-60 or -70 to display flow rate and total, batch controller 33112-52 which provides batchMore Product Information