Masterflex® Gas Mass Flowmeters, Avantor®
Supplier: VWR International
M-50SCCM-D/5M M-200SCCM-D/5M M-500SCCM-D/5M M-50SLPM-D/5M M-100SLPM-D/5M MFLX32908-51
MFLX32908-53EA 3223.2 CAD
MFLX32908-53 MFLX32908-55 MFLX32908-57 MFLX32908-59 MFLX32908-61 MFLX32908-63 MFLX32908-67 MFLX32908-69 MFLX32908-71 MFLX32908-73 MFLX32908-75 MFLX32908-77 MFLX32908-79 MFLX32908-81 MFLX32908-51
Masterflex® Gas Mass Flowmeters, Avantor®
Flow Meters
Measure 130 standard gases—user selectable from display.
  • Accuracy of ±0.8% of reading ±0.2% full-scale; repeatability of ±0.2%
  • 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM
  • Models with 4 to 20 mA output or totalizer are available
These meters measure flow via pressure drop across a laminar flow element (LFE). Because the flow element makes the flow stream laminar, placement in the process does not require straight pipe runs upstream or downstream of the meter, greatly simplifying installation. As compared to thermal massMore Product Information