Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Metric Units, Avantor®
Supplier: KEY INSTRUMENTS, LLC
2510A2L10BVBN-CP 2510A2L11BVBN-CP 2510A2A12BVBN-CP 2510A2A13BVBN-CP 2510A2A14BVBN-CP 2510A2A16BVBN-CP 2510A2A17BVBN-CP 2510A2A18BVBN-CP 2520A4L56BVBN-CP 2520A4L51BVBN-CP 2520A4L50BVBN-CP 2520A4L52BVBN-CP 2520A4L53BVBN-CP 2520A4L54BVBN-CP 2530A4L69BVBN-CP 2530A4L71BVBN-CP 2520A4A40BVBN-CP 2520A4A41BVBN-CP 2520A4A42BVBN-CP 2520A4A43BVBN-CP 2520A4A44BVBN-CP 2520A4A47BVBN-CP 2530A4A72BVBN-CP 2530A4A70BVBN-CP 2540A5L61PV-CP 2540A5A53PV-CP MFLX32466-54 MFLX32466-68 MFLX32466-70
MFLX32460-32EA 200.66 CAD
MFLX32460-32 MFLX32460-34 MFLX32460-40 MFLX32460-42 MFLX32460-44 MFLX32460-46 MFLX32460-48 MFLX32460-50 MFLX32460-52 MFLX32461-30 MFLX32461-32 MFLX32461-34 MFLX32461-36 MFLX32461-38 MFLX32461-40 MFLX32461-42 MFLX32461-44 MFLX32461-46 MFLX32461-50 MFLX32461-52 MFLX32461-54 MFLX32461-56 MFLX32461-58 MFLX32461-60 MFLX32461-62 MFLX32461-64 MFLX32466-56 MFLX32466-66 MFLX32466-54 MFLX32466-68 MFLX32466-70
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Metric Units, Avantor®
Flow Meters
Meter with valve provides flow control through a highly durable meter body.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate
  • Use with air or water for laboratory and industrial applications
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately)More Product Information