Masterflex® Direct-Read Variable Area Flowmeters for Air, Graph-Style, Aluminum, Avantor®
Supplier: BADGER METER INC
HC671A-025-EP HC271A-005-EP HC271A-010-EP HC271A-020-EP HC271A-030-EP HC271A-020-EG HC671A-025-EG HC671A-100-EG HC771A-150-EG
MFLX32206-12EA 1139.18 CAD
MFLX32206-12 MFLX32206-02 MFLX32206-04 MFLX32206-06 MFLX32206-08 MFLX32206-56 MFLX32206-62 MFLX32206-66 MFLX32206-72
Masterflex® Direct-Read Variable Area Flowmeters for Air, Graph-Style, Aluminum, Avantor®
Flow Meters
Easily read graph-style measurements directly.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with air for laboratory and industrial applications
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—More Product Information