Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PMR1-010689 PMR1-013703 PMR1-010692 PMR1-016668 PMR1-010706 PMR1-010713 PMR1-010727 PMR1-010730 PMR1-010735 PMR1-010567 PMR1-010568 PMR1-010570 PMR1-010571 PMR1-010572 PMR1-010579 P13/1-112-19C GF-6500 PMR1-010687 PMR1-010690 PMR1-010694 PMR1-010696 PMR1-010709 PMR1-010710 PMR1-010712 PMR1-016607 PMR1-010714 PMR1-010715 PMR1-010718 PMR1-010731 PMR1-010733 PMR1-010734 GF-1300 P03/1-112-19C PMR1-010584 PMR1-010592 PMR1-010593 PMR1-010594 P23/1-112-19C PMR1-013771 PMR1-010596 PMR1-010597
MFLX32000-04EA 323.9 CAD
MFLX32000-04 MFLX32000-06 MFLX32000-10 MFLX32000-12 MFLX32000-14 MFLX32001-00 MFLX32001-04 MFLX32001-12 MFLX32002-00 MFLX32002-04 MFLX32002-06 MFLX32002-14 MFLX32030-08 MFLX32030-09 MFLX32030-14 MFLX32030-15 MFLX32030-16 MFLX32031-00 MFLX32031-10 MFLX32000-00 MFLX32000-02 MFLX32000-08 MFLX32000-18 MFLX32000-22 MFLX32001-06 MFLX32001-08 MFLX32001-10 MFLX32001-14 MFLX32001-16 MFLX32001-18 MFLX32001-24 MFLX32002-08 MFLX32002-10 MFLX32002-12 MFLX32030-00 MFLX32030-10 MFLX32031-15 MFLX32032-00 MFLX32032-08 MFLX32032-09 MFLX32032-10 MFLX32032-14 MFLX32032-15 MFLX32032-16
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information