Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PMR1-010748 PMR1-010749 PMR1-010751 PMR1-010752 PMR1-010754 GF-8521-2417 PMR1-010755 PMR1-010756 PMR1-010757 PMR1-010769 PMR1-010772 PMR1-010774 PMR1-010775 PMR1-016716 PMR1-010790 PMR1-010791 PMR1-010793 PMR1-010794 PMR1-010796 PMR1-010797 PMR1-010798 PMR1-010804 PMR1-010605 PMR1-010606 PMR1-010608 PMR1-011228 PMR1-010611 PMR1-010618 PMR1-010620 PMR1-010621 PMR1-010624 PMR1-010631 PMR1-010632 PMR1-010633 PMR1-010634 PMR1-010635 PMR1-010637 MFLX32003-18 MFLX32003-20 MFLX32003-24 MFLX32003-26 MFLX32004-16 MFLX32004-26 MFLX32005-04 MFLX32005-18 MFLX32005-20 MFLX32005-24 MFLX32033-09 MFLX32033-15 MFLX32034-08 MFLX32034-14 MFLX32034-15 MFLX32035-15
MFLX32003-00EA 355.5 CAD
MFLX32003-00 MFLX32003-02 MFLX32003-04 MFLX32003-06 MFLX32003-08 MFLX32003-10 MFLX32003-12 MFLX32003-14 MFLX32003-16 MFLX32004-00 MFLX32004-04 MFLX32004-06 MFLX32004-08 MFLX32004-10 MFLX32004-12 MFLX32004-14 MFLX32005-00 MFLX32005-02 MFLX32005-06 MFLX32005-08 MFLX32005-10 MFLX32005-12 MFLX32005-14 MFLX32005-26 MFLX32033-00 MFLX32033-08 MFLX32033-10 MFLX32033-14 MFLX32033-16 MFLX32034-00 MFLX32034-09 MFLX32034-10 MFLX32034-16 MFLX32035-00 MFLX32035-08 MFLX32035-09 MFLX32035-10 MFLX32035-14 MFLX32035-16 MFLX32003-18 MFLX32003-20 MFLX32003-24 MFLX32003-26 MFLX32004-16 MFLX32004-26 MFLX32005-04 MFLX32005-18 MFLX32005-20 MFLX32005-24 MFLX32033-09 MFLX32033-15 MFLX32034-08 MFLX32034-14 MFLX32034-15 MFLX32035-15
Masterflex® Direct-Read Variable-Area Flowmeters with Valve for Air, 150-mm, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information