CPC® DrumQuik® PRO Drum Dispensing System Components
Supplier: VWR International
MFLX31608-20 MFLX31608-21 MFLX31608-22 MFLX31608-25 MFLX31608-38 MFLX31309-67 MFLX31309-65 MFLX31309-66 MFLX31608-31 MFLX31608-35 DQPRO120208 (5PK) DQPRO120712 (5PK) DQPRO202BUT (25PK) DQPRODT0985 (25PK)
MFLX31309-63EA 124.03 CAD
MFLX31309-63 MFLX31608-20 MFLX31608-21 MFLX31608-22 MFLX31608-23 MFLX31608-25 MFLX31608-26 MFLX31608-34 MFLX31608-37 MFLX31608-38 MFLX31309-67 MFLX31309-65 MFLX31309-66 MFLX31608-31 MFLX31608-35 MFLX31608-20BLK MFLX31608-21BLK MFLX31608-22BLK MFLX31608-25BLK MFLX31608-38BLK
CPC® DrumQuik® PRO Drum Dispensing System Components
Fittings and Connectors
Closed-system design ensures the safety of your environment and employees—at all phases of chemical handling.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Indivdual components to keep your dispensing system working smoothly—or build your own
  • Easy-to-use, economical solution for chemical dispensing
  • Durable materials withstand harsh operating conditions
  • Ideal for 5-gallon jerry cans, 55-gallon drums, and IBCs
The CPC® DrumQuik® PRO drum dispensing system offers a simplified solution to your chemical dispensing needs. This systemMore Product Information