CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Supplier: Colder Products
NS6D17006 (10PK) NS4D17004 (25PK) NS6D17008 (10PK) NS6D10008 (10PK) NS4D17006 (25PK) NS4D24004BSPT (25PK) NS6D22008 (10PK) NS4D22006 (25PK) NS6D24008 (10PK) NS4D17002 NS6D42008 NS4D10004BSPT 25PK NS4D22002 25PK NS4D42006 25PK
MFLX31603-22BLKEA 398.16 CAD
MFLX31603-22BLK MFLX31603-23BLK MFLX31603-24BLK MFLX31603-26BLK MFLX31603-31BLK MFLX31603-33BLK MFLX31603-34BLK MFLX31603-35BLK MFLX31603-36BLK MFLX31309-41BLK MFLX31309-49BLK MFLX31309-43BLK MFLX31309-44BLK MFLX31309-47BLK
CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Stop leaks in your line—spillage as low as 0.10 ml.
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation
These CPC® non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. This non-spill series ensures minimal leaks and air inclusions in your process line—protects process purity and minimizes loss due to accidental disconnects. The NS4 and NS6 seriesMore Product Information