CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Supplier: VWR International
MFLX31603-22 MFLX31603-23 MFLX31603-24 MFLX31603-26 MFLX31603-31 MFLX31603-33 MFLX31603-34 MFLX31603-35 MFLX31603-36 MFLX31309-41 MFLX31309-46 MFLX31309-49 MFLX31603-19 MFLX31605-13 MFLX31605-08
MFLX31309-43EA 794.74 CAD
MFLX31309-43 MFLX31309-44 MFLX31309-47 MFLX31603-18 MFLX31603-22 MFLX31603-23 MFLX31603-24 MFLX31603-26 MFLX31603-27 MFLX31603-29 MFLX31603-31 MFLX31603-32 MFLX31603-33 MFLX31603-34 MFLX31603-35 MFLX31603-36 MFLX31603-44 MFLX31309-41 MFLX31309-46 MFLX31309-49 MFLX31603-19 MFLX31605-13 MFLX31605-08
CPC® Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Stop leaks in your line—spillage as low as 0.10 ml.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation
These CPC® non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. This non-spill series ensures minimal leaks and air inclusions in your process line—protects process purity andMore Product Information