Saint Gobain® Sanitary Tri-Clamps to Hose Barb Adapter
Supplier: Saint Gobain Life Sciences
ATP375PPSB (25PK) ATP500PSSB (25PK) ATP375PFSB (25PK) ATP250PPSB (25PK) ATP250PFSB (25PK) ATP250PSSB (25PK) ATP500PPSB (25PK) ATP375PSSB (25PK) ATP500PFSB (25PK)
MFLX31512-01BLKEA 846.01 CAD
MFLX31512-01BLK MFLX31512-08BLK MFLX31512-04BLK MFLX31512-00BLK MFLX31512-03BLK MFLX31512-06BLK MFLX31512-02BLK MFLX31512-07BLK MFLX31512-05BLK
Saint Gobain® Sanitary Tri-Clamps to Hose Barb Adapter
Fittings and Connectors
Quick and secure sanitary tubing attachments are critical in numerous low-pressure laboratory, biotechnology, and pharmaceutical applications. Contamination of fluids and product loss through leakage can be costly. These Sanitary Fittings provide a superior connection to tubing through a single-barb construction, reducing the potential for leakage.
  • Tapered funnel design reduces 'dead space' for exceptionally smooth fluid flow
  • Smooth inner surface prevents particle entrapment
  • Seamless construction prevents contamination by eliminating leaks and areas of
More Product Information