Masterflex® Adapter Fittings, Hose Barb to Female NPT Threaded, Straight, Avantor®
Supplier: VWR International
MFLX31500-40 MFLX31500-41 MFLX31501-40 MFLX31501-41 MFLX31701-85 MFLX31702-20 MFLX31702-22
MFLX31500-40EA 352.34 CAD
MFLX31500-40 MFLX31500-41 MFLX31501-40 MFLX31501-41 MFLX31701-85 MFLX31702-20 MFLX31702-22
Masterflex® Adapter Fittings, Hose Barb to Female NPT Threaded, Straight, Avantor®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery