Union Fittings, Compression Bulkhead, Straight
Supplier: MIDWEST VALVE & FITTING INC
MFLX06390-50 MFLX06390-70 MFLX06390-80 IBD4BU IBD6BU MFLX06390-60 MFLX06390-82 MFLX31406-12 MFLX31406-13 MFLX31406-14 MFLX31406-15
MFLX06390-50EA 29.39 CAD
MFLX06390-50 MFLX06390-70 MFLX06390-80 MFLX31412-13 MFLX31412-14 MFLX06390-60 MFLX06390-82 MFLX31406-12 MFLX31406-13 MFLX31406-14 MFLX31406-15
Union Fittings, Compression Bulkhead, Straight
Fittings and Connectors
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery