CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: VWR International
MFLX31303-40 MFLX31303-41 MFLX31303-42 MFLX31303-43 MFLX31303-45 MFLX31303-46 MFLX31303-47 MFLX31303-48 MFLX31307-40 MFLX31307-41 MFLX31307-45 MFLX31307-46 MFLX31307-48 MFLX31308-74 HFC22612 (10PK) HFC22812 (10PK) HFC221012 (10PK) HFC221212 (10PK) HFCD22612 (10PK) HFCD22812 (10PK) HFCD221012 (10PK) HFCD221212 (10PK) HFC22635 (10PK) HFC22835 (10PK) HFCD22635 (10PK) HFCD22835 (10PK) HFCD221235 (10PK)
MFLX31303-40EA 18.01 CAD
MFLX31303-40 MFLX31303-41 MFLX31303-42 MFLX31303-43 MFLX31303-45 MFLX31303-46 MFLX31303-47 MFLX31303-48 MFLX31307-40 MFLX31307-41 MFLX31307-45 MFLX31307-46 MFLX31307-47 MFLX31307-48 MFLX31308-74 MFLX31303-40BLK MFLX31303-41BLK MFLX31303-42BLK MFLX31303-43BLK MFLX31303-45BLK MFLX31303-46BLK MFLX31303-47BLK MFLX31303-48BLK MFLX31307-40BLK MFLX31307-41BLK MFLX31307-45BLK MFLX31307-46BLK MFLX31307-48BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Inserts
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. ToMore Product Information