CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Supplier: VWR International
MFLX31303-10 MFLX31303-11 MFLX31303-12 MFLX31303-13 MFLX31303-15 MFLX31303-16 MFLX31307-10 MFLX31307-11 MFLX31307-13 MFLX31307-15 MFLX31307-18 HFCD17612 (10PK) HFCD17812 (10PK) HFCD171012 (10PK) HFCD171212 (10PK) HFC17612 (10PK) HFC17812 (10PK) HFCD17635 (10PK) HFCD17835 (10PK) HFCD171235 (10PK) HFC17635 (10PK) HFC171012 HFC17835
MFLX31303-10EA 39.97 CAD
MFLX31303-10 MFLX31303-11 MFLX31303-12 MFLX31303-13 MFLX31303-15 MFLX31303-16 MFLX31303-18 MFLX31307-10 MFLX31307-11 MFLX31307-13 MFLX31307-15 MFLX31308-34 MFLX31308-63 MFLX31307-18 MFLX31303-10BLK MFLX31303-11BLK MFLX31303-12BLK MFLX31303-13BLK MFLX31303-15BLK MFLX31303-16BLK MFLX31307-10BLK MFLX31307-11BLK MFLX31307-13BLK MFLX31307-15BLK MFLX31308-34BLK MFLX31308-63BLK
CPC® High-Flow Quick-Disconnect Fittings, Hose Barb Bodies
Fittings and Connectors
No more leaks—valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many 1/2" flow couplings in a 3/8” body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. ToMore Product Information