Masterflex® Single-Use Filling Needles
Supplier: VWR International
MFLX30719-00 MFLX30719-02 MFLX30719-04 MFLX30719-06
MFLX30719-00EA 497.7 CAD
MFLX30719-00 MFLX30719-02 MFLX30719-04 MFLX30719-06
Masterflex® Single-Use Filling Needles
Fittings and Connectors
A high-quality engineered solution for every application in the fill-finish workflow.
  • Material compliant to USP Class VI, FDA 21 CFR, ADCF
  • Double bagged in ISO Class 7 cleanroom
  • High purity and contamination-free materials
  • Ready to use; increases productivity
  • Straightness <2 mm/M for consistent uniformity
  • Oil-free and adhesive-free construction
  • Repeatable performance with high-speed and accuracy
Filling speed, precision and accuracy are at the heart of the design process. Masterflex® Single Use Filling Needles are a cost‐effective and uniqueMore Product Information