Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Supplier: VWR International
MFLX30544-20
MFLX30544-20EA 289.14 CAD
MFLX30544-20
Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Seals Gaskets
Easily sample fluid in critical high-purity system.
  • Three internal ports accept thermocouple probe0
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.