CPC® MicroCNX™ Series Genderless Aseptic Fitting
Supplier: Colder Products
CNX17001HT CNX17002HT CNX17003HT
MFLX20202-00EA 1035.48 CAD
MFLX20202-00 MFLX20202-01 MFLX20202-02
CPC® MicroCNX™ Series Genderless Aseptic Fitting
Fittings and Connectors
Provide easy, secure sterile connections even in nonsterile environments now in newly designed smaller sizes.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
  • Sterile micro-connectors eliminates the need to purchase, calibrate, validate, and maintain tube welding equiptment
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use andMore Product Information