Value Plastics Luer Fittings and Adapters
Supplier: VWR International
MFLX15973-17 MFLX15973-45 MFLX30651-72 MFLX15955-84 MFLX15955-87 MFLX15955-95 MFLX15973-12 MFLX15955-81 MFLX15955-97 MFLX15973-25 MFLX15973-26 MFLX15955-79 MFLX15955-83 ABSML-6005-11000
MFLX15955-78EA 35.55 CAD
MFLX15955-78 MFLX15955-82 MFLX15955-93 MFLX15955-96 MFLX15955-98 MFLX15956-07 MFLX15956-08 MFLX15956-09 MFLX15973-11 MFLX15973-14 MFLX15973-15 MFLX15973-17 MFLX15973-18 MFLX15973-20 MFLX15973-29 MFLX15973-45 MFLX30495-72 MFLX30651-72 MFLX15955-84 MFLX15955-87 MFLX15955-95 MFLX15973-12 MFLX15955-81 MFLX15955-97 MFLX15973-25 MFLX15973-26 MFLX15955-79 MFLX15955-83 MFLX15973-45BLK
Value Plastics Luer Fittings and Adapters
Fittings and Connectors
Premium luer fittings for critical applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. ChooseMore Product Information