ODE NSF-Certified Two-Way Solenoid Valves
Supplier: FLUID CONVEYING PRODUCTS
21JP1R1V23-T0 21JPAR1V23-T0
MFLX08617-94CS 47.7 CAD
MFLX08617-94 MFLX08617-96
ODE NSF-Certified Two-Way Solenoid Valves
Valves Solenoid Valves
Wetted materials fully compliant with NSF standards to ensure water and food safety.
  • Valves create zero differential pressure
  • Integrated mounting holes for ease of installation or replacement
  • Low actuation wattage required, resulting in low energy consumption and operating cost
These two-way normally closed solenoid valves are used in sterilization, process, and vending of liquids, beverages, and food where NSF certification is required. High-quality virgin materials provide extended service life, and high pressure and temperature resistance TheMore Product Information