Masterflex® L/S® Portable Sampling Pumps, Avantor®
Supplier: Avantor Fluid Handling
7571-02
MFLX07571-02EA 3981.6 CAD
MFLX07571-02
Masterflex® L/S® Portable Sampling Pumps, Avantor®
Pumps Drum/Transfer Pumps
Sample depths down to 7.9 meters (26 feet).
  • Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switch
  • Reversible flow and prime/purge function
  • Includes a Masterflex® L/S® Easy-Load pump head
  • Also accepts Masterflex® L/S® Standard pump heads (order separately)
  • Floats up to 30 minutes if dropped in water when case is closed and latched
  • IP54-rated control panel protects against inclement weather and splashing water
  • Rechargeable battery — ideal for field applications
The Masterflex® L/S® portableMore Product Information