Masterflex® L/S® Analog Variable-Speed Console Drives, Avantor®
Supplier: Avantor Fluid Handling
7555-00 7555-05 7555-10 7555-15
MFLX07555-00EA 2133 CAD
MFLX07555-00 MFLX07555-05 MFLX07555-10 MFLX07555-15
Masterflex® L/S® Analog Variable-Speed Console Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Compact and cost effective for general fluid transfer applications.
  • Flow rate 0.42 to 2900 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Drives accept nine different pump head types: Standard, Easy-Load®,More Product Information