Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
7019-00 7019-25 7019-26 77601-62 77602-02 77602-12
MFLX07019-00EA 1207.12 CAD
MFLX07019-00 MFLX07019-25 MFLX07019-26 MFLX77601-62 MFLX77602-02 MFLX77602-12
Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Designed and optimized for one tubing size
  • Three roller design for smooth flow
  • Choose from stainless steel (SS) or cold-rolled steel (CRS) rotor / roller for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
Standard heads are precision molded to accommodate a single size of tubing—use for common fluid transfer applications requiring accuracy and excellent repeatability. AvailableMore Product Information