Masterflex® L/S® Standard Long-Shaft Pump Head Rotor Assembly, Avantor®
Supplier: Avantor Fluid Handling
7015-76
MFLX07015-76EA 469.26 CAD
MFLX07015-76
Masterflex® L/S® Standard Long-Shaft Pump Head Rotor Assembly, Avantor®
Pumps Peristaltic Pumps
Get the most out of your Masterflex® L/S® Standard Pump Head with these replacement parts.
  • Rotor is constructed of stainless steel (SS) for enhanced durability
Replacement parts to keep your pump head operating even longer.