Masterflex® Ismatec® Microbore Pump Tubing, Tygon® E-Lab, Avantor®
Supplier: VWR International
MFLX06460-09 MFLX06460-10 MFLX06460-12 MFLX06460-14 MFLX06460-16 MFLX06460-18 MFLX06460-19 MFLX06460-22 MFLX06460-24 MFLX06460-26 MFLX06460-27 MFLX06460-28 MFLX06460-30 MFLX06460-31 MFLX06460-32 MFLX06460-34 MFLX06460-36 MFLX06460-38 MFLX06460-39 MFLX06460-40 MFLX06460-42 MFLX06460-44 MFLX06460-46 MFLX06460-48 MFLX06460-49 MFLX06460-29
MFLX06460-09EA 295.46 CAD
MFLX06460-09 MFLX06460-10 MFLX06460-12 MFLX06460-14 MFLX06460-16 MFLX06460-18 MFLX06460-19 MFLX06460-22 MFLX06460-24 MFLX06460-26 MFLX06460-27 MFLX06460-28 MFLX06460-30 MFLX06460-31 MFLX06460-32 MFLX06460-34 MFLX06460-36 MFLX06460-38 MFLX06460-39 MFLX06460-40 MFLX06460-42 MFLX06460-44 MFLX06460-46 MFLX06460-48 MFLX06460-49 MFLX06460-29
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® E-Lab, Avantor®
Tubing Peristaltic Pump Tubing
Ideal for general transfer applications.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: General laboratory use
Characteristics: Nontoxic, nonaging, nonoxidizing, easy flow monitoring, transparent
The overall length of E-Lab 06460-seriesMore Product Information