Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Supplier: VWR International
MFLX06456-13 MFLX06456-14 MFLX06456-16
MFLX06456-13EA 161.16 CAD
MFLX06456-13 MFLX06456-14 MFLX06456-16
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Avantor®
Tubing Peristaltic Pump Tubing
Ultra low spallation protects fluid purity – ideal for sensitive and costly fluids and for filtration applications.
  • Genuine Masterflex® pump tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Gentle, low pulsation and accurate dosing and flow when used in the Masterflex Ultrapharm™ L/S® pump head
  • Long life under
More Product Information