Masterflex® L/S® 2-Stop Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
6421-13 6421-14 6421-15 6421-16 6421-24 6421-25 6421-35
MFLX06421-13EA 178.54 CAD
MFLX06421-13 MFLX06421-14 MFLX06421-15 MFLX06421-16 MFLX06421-24 MFLX06421-25 MFLX06421-35
Masterflex® L/S® 2-Stop Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases and
More Product Information