Masterflex® L/S® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Supplier: VWR International
MFLX06401-13 MFLX06401-14 MFLX06401-16 MFLX06401-17 MFLX06401-18 MFLX06401-25
MFLX06401-13EA 133.67 CAD
MFLX06401-13 MFLX06401-14 MFLX06401-16 MFLX06401-17 MFLX06401-18 MFLX06401-25
Masterflex® L/S® Precision Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
Ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
More Product Information