Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Supplier: VWR International
5005-1000 5005-1200 5005-7000
MFLX06040-45EA 149.47 CAD
MFLX06040-45 MFLX06040-46 MFLX06040-47 MFLX06040-49 MFLX06040-51 MFLX06040-52 MFLX06040-53 MFLX06040-29
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Pumps Drum/Transfer Pumps
Dispense liquids safely and quickly from most storage containers.
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
These hand-operated pumps feature a universal soft PVC bung consisting of a conical center and two conical sleeves allowing for an airtight seal between the pump and most liquid storage container openings. The telescopic immersion tube adapts itself to the container height on insertion, ensuring good removal of residual liquid. Various tube diameters areMore Product Information