Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
T51/1-N044-40S P01/1-N044-40G P01/1-N044-40ST P01/1-N044-40C T51/1-044-40G P21/1-N044-40G P21/1-N044-4OC MFLX03229-35 MFLX03295-32 MFLX03295-34 MFLX03295-36
MFLX03217-78EA 786.84 CAD
MFLX03217-78 MFLX03227-32 MFLX03227-34 MFLX03227-36 MFLX03227-87 MFLX03229-33 MFLX03229-37 MFLX03229-35 MFLX03295-32 MFLX03295-34 MFLX03295-36
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information