Masterflex® Correlated Variable-Area Flowmeters with Valve, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P04/1-N032-41G P04/1-N032-41ST P04/1-N062-01G P04/1-N062-01ST P04/1-N112-02G P04/1-N082-03G P04/1-N082-03ST P04/1-N092-04G P04/1-N092-04ST P04/1-N102-05G P04/1-N102-05ST GF-8521-1606 P04/1-N034-39G T54/1-N032-41G T54/1-N032-41S T54/1-N062-01G T54/1-N062-01S T54/1-N112-02G T54/1-092-04G T54/1-034-39G T54/1-N034-39S P24/1-N032-41G P24/1-N032-41ST P24/1-N062-01G P24/1-N062-01ST P24/1-N112-02G P24/1-N112-02ST P24/1-N082-03G P24/1-N082-03ST P24/1-N092-04G P24/1-N092-04ST P24/1-N102-05G P24/1-N034-39G P24/1-N034-39ST P14/1-N032-41G P14/1-N032-41ST P14/1-N062-01G P14/1-N062-01ST P14/1-N112-02G P14/1-N112-02ST P14/1-N102-05G P14/1-N102-05ST MFLX03217-14 MFLX03217-30 MFLX03217-65 MFLX03217-66 MFLX03217-70 MFLX03217-75 MFLX03217-77 MFLX03217-80 MFLX03219-01 MFLX03219-03 MFLX03219-27 MFLX03294-16 MFLX03294-18 MFLX03294-30
MFLX03217-00EA 338.12 CAD
MFLX03217-00 MFLX03217-02 MFLX03217-04 MFLX03217-06 MFLX03217-08 MFLX03217-10 MFLX03217-12 MFLX03217-16 MFLX03217-18 MFLX03217-20 MFLX03217-22 MFLX03217-24 MFLX03217-26 MFLX03217-28 MFLX03217-50 MFLX03217-51 MFLX03217-55 MFLX03217-56 MFLX03217-60 MFLX03217-61 MFLX03217-71 MFLX03217-81 MFLX03217-85 MFLX03219-05 MFLX03219-07 MFLX03219-09 MFLX03219-11 MFLX03219-13 MFLX03219-15 MFLX03219-17 MFLX03219-19 MFLX03219-21 MFLX03219-23 MFLX03219-25 MFLX03219-29 MFLX03219-31 MFLX03294-00 MFLX03294-04 MFLX03294-06 MFLX03294-08 MFLX03294-10 MFLX03294-12 MFLX03294-14 MFLX03294-24 MFLX03294-26 MFLX03217-14 MFLX03217-30 MFLX03217-65 MFLX03217-66 MFLX03217-70 MFLX03217-75 MFLX03217-77 MFLX03217-80 MFLX03219-01 MFLX03219-03 MFLX03219-27 MFLX03294-16 MFLX03294-18 MFLX03294-30
Masterflex® Correlated Variable-Area Flowmeters with Valve, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information