Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
2P04/6C 2P04MT/6C 2P24/6C 2P24MT/6C 4P04MT/6C 5P04/6C 2P04/1C 2P04MT/1C 2P24/1C 2P24MT/1C 3P04MT/1C 3P24/1C 3P24MT/1C 4P04MT/1C 4P24/1C 4P24MT/1C 5P24MT/1C 6P04MT/1C 6P24/1C MFLX03214-28 MFLX03214-30 MFLX03214-36 MFLX03214-46 MFLX03214-52 MFLX03214-56 MFLX03214-64 MFLX03214-66 MFLX03214-68 MFLX03214-74 MFLX03214-76 MFLX03214-78 MFLX03214-80 MFLX03215-28 MFLX03215-46 MFLX03215-64 MFLX03215-74 MFLX03215-80
MFLX03214-10EA 243.32 CAD
MFLX03214-10 MFLX03214-12 MFLX03214-16 MFLX03214-18 MFLX03214-50 MFLX03214-62 MFLX03215-10 MFLX03215-12 MFLX03215-16 MFLX03215-18 MFLX03215-30 MFLX03215-34 MFLX03215-36 MFLX03215-50 MFLX03215-52 MFLX03215-56 MFLX03215-68 MFLX03215-76 MFLX03215-78 MFLX03214-28 MFLX03214-30 MFLX03214-36 MFLX03214-46 MFLX03214-52 MFLX03214-56 MFLX03214-64 MFLX03214-66 MFLX03214-68 MFLX03214-74 MFLX03214-76 MFLX03214-78 MFLX03214-80 MFLX03215-28 MFLX03215-46 MFLX03215-64 MFLX03215-74 MFLX03215-80
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Flow Meters
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless
More Product Information