George Fischer Type 546 Ball Valves 42387
Supplier: BURT PROCESS EQUIPMENT
161.546.341 161.546.342 161.546.343 161.546.344 163.546.351 163.546.352 175.546.332(TYPE-546) MFLX01371-10 MFLX01374-94
MFLX01357-01EA 183.28 CAD
MFLX01357-01 MFLX01357-03 MFLX01357-05 MFLX01357-07 MFLX01371-08 MFLX01371-09 MFLX01374-92 MFLX01371-10 MFLX01374-94
George Fischer Type 546 Ball Valves 42387
Valves Ball Valves
Optimize safety with this true union ball valve.
  • True union ball valve with added safety and functionality for a diverse range of applications
  • PTFE ball seat with high abrasion resistance
  • Built-in tool for easy adjustment of ball torque
  • Threaded mounting inserts for fixed-point attachment
Ball valves are designed primarily for on/off service. These valves contain a ball with a hole through it. A handle or electronic actuator rotates the ball 90 degrees, turning the flow on or off.