KONTES® Fritted Hirsch Funnels, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
955250-3523 955250-5023 955250-5026 955250-5044 955250-7523 955250-7526 955250-7544
KT955250-3523CS 100.87 CAD
KT955250-3523 KT955250-5023 KT955250-5026 KT955250-5044 KT955250-7523 KT955250-7526 KT955250-7544
KONTES® Fritted Hirsch Funnels, KIMBLE®, DWK Life Sciences
Funnels Filtration Funnels Filter Funnels
Funnels feature a conical shape that directs filtrate through a reduced-area frit to concentrate crystal residue for rewashing.