Stir Blades, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
789030-0019 789030-0020 789030-0021 789030-0022 789030-0023 789030-0024
46000-220EA 71.97 CAD
46000-220 KT789030-0020 KT789030-0021 KT789030-0022 KT789030-0023 KT789030-0024
Stir Blades, Kimble Chase
Stirrers Stirring Blades
PTFE stirrer blades fit Kimble Chase shafts 786500 and 786520
The blade is 3.2mm (1/8") thick. A 10.2cm (4") cylindrical reaction flask accepts PTFE blade 789030-0022.
Order Now
SPECIFICATIONS
Environmentally Preferable