KIMAX® Volumetric Flasks with Screw Cap, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
621600-0005 621600-0025 621600-0050 621600-0250 621600-2000 621600-0010 621600-0100 621600-0500 621600-1000
KT621600-0005EA 80.42 CAD
KT621600-0005 KT621600-0025 KT621600-0050 KT621600-0250 KT621600-2000 KT621600-0010 KT621600-0100 KT621600-0500 KT621600-1000
KIMAX® Volumetric Flasks with Screw Cap, Class A, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Threaded flasks with a PTFE/rubber-lined phenolic cap.
Order Now
SPECIFICATIONS
Environmentally Preferable