KONTES® Kuderna-Danish Concentrators, Ungraduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
570010-0250
KT570010-0250EA 814.83 CAD
KT570010-0250
KONTES® Kuderna-Danish Concentrators, Ungraduated, Kimble Chase
Concentrators
Developed for the concentration of trace amounts of sample dissolved in organic solvents
Preparation involves filling the flask between 40 to 60% of capacity. To prevent sample loss initially, the column should be pre-wet with about 1ml of the solvent used in the concentration. Column is designed to speed evaporation with reduced hold-up.
Charged assembly should be placed over a vigorously boiling water bath. Water level should be maintained just below the lower [ST] joint and the apparatus mounted so that the lower rounded surface of the flask is bathed in steam.
Note: If solventMore Product Information