KIMBLE® Snyder Distillation Column, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
503100-0003
KT503100-0003EA 546.76 CAD
KT503100-0003
KIMBLE® Snyder Distillation Column, DWK Life Sciences
Distillation Components Distillation Columns
Improved design speeds evaporation and reduces hold-up within the column.
  • Recoveries are excellent, example with pesticides such as lindane and parathion in petroleum ether or ethyl ether
  • Higher boiling solvents also may be used to good advantage
  • Each chamber is approximately 50 mm long
  • Manufactured from 33 expansion, low extractable borosilicate glass
Kimble® improved snyder distillation column with 3-chamber and 24/40 [ST] joints.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.