TLC Reagent Sprayers with Screw Thread Ground Joints, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
422530-0010 422530-0125 422530-0250 422530-0500 422531-0050 422531-0125 422531-0250 422531-0500 422532-0010 422532-0050 422532-0125 422532-0250 422533-0019 422534-0019 422530-0025
KT422530-0010EA 711.95 CAD
KT422530-0010 KT422530-0125 KT422530-0250 KT422530-0500 KT422531-0050 KT422531-0125 KT422531-0250 KT422531-0500 KT422532-0010 KT422532-0050 KT422532-0125 KT422532-0250 KT422532-0500 KT422533-0019 KT422534-0019 KT422530-0025
TLC Reagent Sprayers with Screw Thread Ground Joints, Kimble Chase
Atomizers
These sprayers are manufactured from 33 expansion, low extractable borosilicate glass, conforming to USP Type I and ASTM E438, Type I, Class A requirements.
These reagent sprayers have a screw thread joint for connecting the sprayer head to the flask. The joint features a connecting cap and loosening ring for easy disconnecting in the event of reagent crystallization or residue formation. These sprayers will operate on as little as 1 psi air pressure, with optimal results at 3 to 5 psi.
Caution: The connecting cap, loosening ring and o-ring should be removed before autoclaving.