KONTES® FLEX-COLUMN® Chromatography Columns, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420400-0720 420400-1005 420400-1510 420400-1515 420400-1520 420400-1575 420400-2511 420400-2512 420400-2530 420400-2550 420400-2575 420401-1050 420401-2550 420400-0704 420400-0705 420400-0710 420400-0715 420400-0730 420400-0750 420400-1010 420400-1012 420400-1015 420400-1030 420400-1050 420400-1505 420400-1511 420400-1512 420400-1517 420400-1530 420400-1550 420400-2505 420400-2510 420400-2515 420400-2520 420401-0704 420401-0705 420401-0710 420401-0715 420401-0720 420401-0730 420401-0750 420401-1005 420401-1010 420401-1011 420401-1012 420401-1020 420401-1030 420401-1505 420401-1510 420401-1511 420401-1512 420401-1515 420401-1517 420401-1520 420401-1530 420401-1550 420401-1575 420401-2505 420401-2510 420401-2511 420401-2512 420401-2515 420401-2520 420401-2530 420401-2575
KT420400-0720PK 577.63 CAD
KT420400-0720 KT420400-1005 KT420400-1510 KT420400-1515 KT420400-1520 KT420400-1575 KT420400-2511 KT420400-2512 KT420400-2530 KT420400-2550 KT420400-2575 KT420401-1050 KT420401-2550 KT420400-0704 KT420400-0705 KT420400-0710 KT420400-0715 KT420400-0730 KT420400-0750 KT420400-1010 KT420400-1012 KT420400-1015 KT420400-1030 KT420400-1050 KT420400-1505 KT420400-1511 KT420400-1512 KT420400-1517 KT420400-1530 KT420400-1550 KT420400-2505 KT420400-2510 KT420400-2515 KT420400-2520 KT420401-0704 KT420401-0705 KT420401-0710 KT420401-0715 KT420401-0720 KT420401-0730 KT420401-0750 KT420401-1005 KT420401-1010 KT420401-1011 KT420401-1012 KT420401-1020 KT420401-1030 KT420401-1505 KT420401-1510 KT420401-1511 KT420401-1512 KT420401-1515 KT420401-1517 KT420401-1520 KT420401-1530 KT420401-1550 KT420401-1575 KT420401-2505 KT420401-2510 KT420401-2511 KT420401-2512 KT420401-2515 KT420401-2520 KT420401-2530 KT420401-2575
KONTES® FLEX-COLUMN® Chromatography Columns, Kimble Chase
Chromatography Columns
These economical columns are constructed with borosilicate glass barrels, molded polypropylene end caps, and polyethylene bed supports.
Polypropylene packing reservoir funnels are for use in initial column packing or as small buffer reservoirs. Glass packing reservoirs are ideal for packing gel slurries into clumns or acting as a buffer reservoir in gravity flow chromatography. Glass reservoirs include a polypropylene upper cap with male Luer-lock fitting.
Flow adapters allow easy adjustment of column bed volume and offer improved column performance and chromatogram resolution. FlowMore Product Information