Alcohol, anhydrous, Reagent Grade for histology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Reagent alcohol

Management of Change (MOC) category = N

A478-09
JTA478-9CS 1066.01 CAD
JTA478-9 CAJTA478-9
Alcohol, anhydrous, Reagent Grade for histology, J.T.Baker®
Alcohol

(Made from Specially Denatured Alcohol 3A)


Intended for nonfuel end purposes only.

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Color (APHA) ≤10
Specific Gravity at 25°C 0.785 - 0.795
Residue after Evaporation ≤0.0100 %
Water (H₂O) ≤0.5 %
Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.
For General Purpose Reagent Use Only.
NOTE: Intended for nonfuel end purposes only.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR