Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

Management of Change (MOC) category = N

BAKR9508-15 9508-02 9508-05 9508-01 9508-03
CAJT9508-15EA 6014.43 CAD
CAJT9508-15 JT9508-2 JT9508-5 JT9508-1 JT9508-3 CAJT9508-69
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Acetic acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.


Caution: IMPORTANT: Material will freeze if stored below 17 °C (63 °F).

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (by GC, corrected for water) ≥99.7 %
Color (APHA) ≤10
Dilution Test Passes Test
Residue after Evaporation ≤0.001 %
Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
Chloride (Cl) ≤1 ppm
Sulfate (SO₄) ≤1 ppm
Trace Impurities - Iron (Fe) ≤200.0 ppb
Substances Reducing Dichromate Passes Test
Substances Reducing Permanganate Passes Test
Titrable Base (meq/g) ≤0.0004
Acetaldehyde ≤0.005 %
Sensitivity Passes Test
Trace Impurities - Arsenic and Antimony (as As) ≤50.0 ppb
Trace Impurities - Boron (B) ≤100.0 ppb
Trace Impurities - Chromium (Cr) ≤200.0 ppb
Trace Impurities - Cobalt (Co) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Heavy Metals ≤0.5 ppm
Trace Impurities - Lead (Pb) ≤300.0 ppb
Trace Impurities - Manganese (Mn) ≤200.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤300.0 ppb
Trace Impurities - Zinc (Zn) ≤200.0 ppb
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR