N-Methyl-2-pyrrolidone (NMP) ≥99.5%, ALEG® for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

Management of Change (MOC) category = R

6397-05
JT6397-5CS 9286.8 CAD
JT6397-5 CAJT6397-5
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, ALEG® for microelectronic, J.T.Baker®
N-Methyl-2-pyrrolidone (NMP)

Positive photoresist stripper (NMP).

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay (C₅H₉NO) ≥99.5 %
Color (APHA) ≤20
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Free Amines (as CH₃NH₂) ≤50 ppm
Chloride (Cl) ≤0.5 ppm
Phosphate (PO₄) ≤1 ppm
Trace Impurities - Aluminum (Al) ≤100.0 ppb
Arsenic and Antimony (as As) ≤100.0 ppb
Trace Impurities - Barium (Ba) ≤100.0 ppb
Trace Impurities - Boron (B) ≤100.0 ppb
Trace Impurities - Calcium (Ca) ≤300 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Cobalt (Co) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤100.0 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Lithium (Li) ≤100.0 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Potassium (K) ≤100 ppb
Trace Impurities - Silver (Ag) ≤100.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Strontium (Sr) ≤100.0 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Vanadium (V) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤100.0 ppb
Particle Count - 1.0 µm and greater (Rion KS42AF) ≤10 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR