Sodium thiosulfate pentahydrate 99.5-101.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR3946-01 3946-05 3946-07
CAJT3946-7EA 2061.54 CAD
CAJT3946-7 CAJT3946-1 CAJT3946-5 JT3946-5 JT3946-7 CAJT3946-69
Sodium thiosulfate pentahydrate 99.5-101.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂S₂O₃·5H₂O) (by iodometry) 99.5 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.0 - 8.4
Nitrogen Compounds (as N) ≤0.002 %
Sulfate and Sulfite (as SO₄) ≤0.1 %
Sulfide (S) Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR