I-CHEM™ Wide Mouth Jars with Caps, Thermo Scientific
Supplier: Thermo Scientific
120-0060 120-0125 120-0250 120-2000 121-0125 121-1000 121-4000 140-0120 220-0060 220-0125 220-0250 220-0500 220-2000 221-0125 221-0500 221-1000 221-2000 221-4000 240-0060 240-0120 320-0060 320-0125 320-0250 320-0500 320-2000 321-0125 321-0250 321-0500 321-1000 321-2000 321-4000 340-0060 340-0120 V220-0125 V221-1000 V320-0125 V320-0500 V321-1000 121-0250 221-0250 340-0250
IR120-0060CS 243.01 CAD
IR120-0060 IR120-0125 IR120-0250 IR120-0500 IR120-2000 IR121-0125 IR121-0500 IR121-1000 IR121-4000 IR140-0060 IR140-0120 IR220-0060 IR220-0125 IR220-0250 IR220-0500 IR220-2000 IR221-0125 IR221-0500 IR221-1000 IR221-2000 IR221-4000 IR240-0060 IR240-0120 IR320-0060 IR320-0125 IR320-0250 IR320-0500 IR320-2000 IR321-0125 IR321-0250 IR321-0500 IR321-1000 IR321-2000 IR321-4000 IR340-0060 IR340-0120 IRV220-0125 IRV220-0250 IRV220-0500 IRV221-0125 IRV221-1000 IRV320-0125 IRV320-0250 IRV320-0500 IRV321-0250 IRV321-1000 IR121-0250 IR121-2000 IR221-0250 IRV321-0125 IR-340-0250
I-CHEM™ Wide Mouth Jars with Caps, Thermo Scientific
Bottles Water Sampling Bottles
Glass jars for use in soil, sludge, and sediment sampling.
I-CHEM Certified™ 200 and 300 series bottles are processed to meet or exceed US EPA Specifications and Guidance for Contaminent-Free Sample Containers. I-CHEM Certified™ 300 series bottles include a Certificate of Analysis with a bar-coded production number in every case. Each bottle in the 300 series has a bar-code label for absolute traceability and is for use with automated sample tracking systems.
I-CHEM™ 100 Series bottles, for general use, are not processed or certified.
Select sizes are for use in volatile organicMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable