I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Supplier: Thermo Scientific
311-0500 311-1000 312-0250 312-0500 312-0950 313-0250 313-0500 313-1000 319-0125 319-1000 312-4000 313-0125
IR311-0500CS 374.76 CAD
IR311-0500 IR311-1000 IR312-0250 IR312-0500 IR312-0950 IR313-0250 IR313-0500 IR313-1000 IR319-0125 IR319-1000 IR312-4000 IR313-0125
I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
HDPE containers, white, with LDPE-lined polypropylene cap, narrow/wide neck. Suitable for metals analyses and water quality parameters.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy filling and sample retrieval
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
I-CHEM Certified™ 300 series containers are processed to meet or exceed 'USEPA Specifications and Guidance for Contaminent-Free Containers'.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable