Anti-C4 Goat Polyclonal Antibody (Fluorescein)

Supplier: MP Biomedicals
0855168
IC0855168EA 497.98 CAD
IC0855168
Anti-C4 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Product is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat IgG fraction to human complement C4 and buffer salts.

Type: Primary 
Antigen: C4
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR