Ionomycin free acid ≥98% (by HPLC, TLC), waxy colorless solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0218382301 0218382305
IC0218382301EA 343.62 CAD
IC0218382301 IC0218382305
Ionomycin free acid ≥98% (by HPLC, TLC), waxy colorless solid
Ionomycin

Antibiotic. Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations at efficiencies that are greater than or equal to those for Ca2+. Apoptosis inducer. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase and induces central demyelination. Used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2 and IL-4 usually in conjunction with PMA. ADAM10 agonist. Potent inducer of shedding CXCL16. TREK-1 channels inhibitor. PPARgamma ligand with a unique binding mode. Shows effective glucose-lowering activity in a mouse model of diabetes.


  • Purity : >98% (HPLC, TLC)
  • Product Type : Biochemicals
  • Host : Isolated from Streptomyces conglobatus.
  • Solubility : Soluble in methanol or DMSO.

Formula: C₄₁H₇₂O₉
MW: 709.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06798385
CAS Number: 56092-81-0

Order Now

Specification Test Results

Identity Test Passes
Purity >98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR