Piceatannol ≥98% (by HPLC), crystalline solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

0218335005 0218335025
IC0218335005EA 673.55 CAD
IC0218335005 IC0218335025
Piceatannol ≥98% (by HPLC), crystalline solid
Piceatannol

Wide range of tyrosine and serine/threonine protein kinase inhibitor, including Syk, p56lck, PKA, PKC, MLCK, CDPK, JNK and PI3K. Inhibits the tyrosine phosphorylation of STAT3 and STAT5. Potent apoptosis inducer. Potent anticancer compound.


  • Purity : >98% (HPLC)
  • Product Type : Biochemicals
  • Host : Synthetic. Originally isolated from Euphorbia lagascae


Suppresses NF-kB activation through IkBalpha kinase inhibition. Activator of human deacetylase SIRT1 (sirtuin 1). Potent antioxidant with anti-proliferative, anti-inflammatory and cardioprotective properties. Neuoprotective. Adipogenesis inhibitor. Promotes glucose uptake, AMPK phosphorylation and GLUT4 translocation. Autophagy inducer


Solubility : Soluble in DMSO, dimethyl formamide, ethanol, methanol, acetone or ethyl acetate. Slightly soluble in water.

Formula: C₁₄H₁₂O₄
MW: 244.2427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00221715
CAS Number: 10083-24-6

Order Now

Specification Test Results

Identity Test Passes
Purity 98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR